Merck
  • Biosynthesis of polyhydroxyalkanoates containing 2-hydroxy-4-methylvalerate and 2-hydroxy-3-phenylpropionate units from a related or unrelated carbon source.
Biosynthesis of polyhydroxyalkanoates containing 2-hydroxy-4-methylvalerate and 2-hydroxy-3-phenylpropionate units from a related or unrelated carbon source.
Journal of bioscience and bioengineering (2017-11-28)
Shoji Mizuno, Yukio Enda, Azusa Saika, Ayaka Hiroe, Takeharu Tsuge
ZUSAMMENFASSUNG
The discovery of the lactate-polymerizing enzyme (LPE) enabled the biosynthesis of a polyhydroxyalkanoate (PHA) containing 2-hydroxyalkanoate (2HA). Amino acids are potential precursors of 2HA with various side chain structures if appropriate enzymes are used to convert amino acids to 2HA-coenzyme A (CoA) as the substrate for LPE. In this study, the suitability and utility of (R)-2-hydroxy-4-methylvalerate (2H4MV) dehydrogenase (LdhA) and 2H4MV-CoA transferase (HadA) from Clostridium difficile as 2HA-CoA-supplying enzymes were investigated. By expressing LPE, LdhA, and HadA in Escherichia coli DH5α, we successfully produced poly(3-hydroxybutyrate-co-2HA) [P(3HB-co-2HA)] from a related or unrelated carbon source. The 2HA units incorporated into PHA from unrelated carbon sources were primarily 2H4MV and 2-hydroxy-3-phenylpropionate (2H3PhP), which were assumed to be derived from endogenous leucine and phenylalanine, respectively. Furthermore, P(3HB-co-22 mol% 2HA) synthesis was demonstrated by means of saccharified sugars, which are an abundant and renewable feedstock for polymer production from hemicellulosic biomass (Japanese cedar) as the carbon source. Our study shows that several types of 2HA units such as 2H4MV and 2H3PhP are endogenous monomers for PHA biosynthesis in E. coli expressing LdhA and HadA.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
6-Phosphonohexansäure, 97%