Merck
IL-17-driven intestinal fibrosis is inhibited by Itch-mediated ubiquitination of HIC-5.
Mucosal immunology (2017-06-15)
J Paul, A K Singh, M Kathania, T L Elviche, M Zeng, V Basrur, A L Theiss, K Venuprasad
ZUSAMMENFASSUNG
Intestinal fibrosis is a major complication in inflammatory bowel diseases, but the regulatory mechanism that inhibits fibrosis remains unclear. Here we demonstrate that Itch-/-myofibroblasts express increased amounts of profibrotic collagen type I and α-SMA in response to IL-17. Mechanistically, we demonstrate that Itch directly binds to HIC-5 and targets it for K63-linked ubiquitination to inhibit IL-17-driven intestinal fibrosis. Reconstitution of Itch-/- myofibroblasts with wild-type Itch but not the Itch-C830A mutant normalized the expression of profibrotic genes. Similarly, shRNA-mediated inhibition of HIC-5 normalized the expression of profibrotic gene expression. Thus, we have uncovered a novel mechanism by which Itch negatively regulates intestinal fibrosis.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
MISSION® esiRNA, targeting human TGFB1I1