Merck
  • Manganese Increases the Sensitivity of the cGAS-STING Pathway for Double-Stranded DNA and Is Required for the Host Defense against DNA Viruses.
Manganese Increases the Sensitivity of the cGAS-STING Pathway for Double-Stranded DNA and Is Required for the Host Defense against DNA Viruses.
Immunity (2018-04-15)
Chenguang Wang, Yukun Guan, Mengze Lv, Rui Zhang, Zhaoying Guo, Xiaoming Wei, Xiaoxia Du, Jing Yang, Tong Li, Yi Wan, Xiaodong Su, Xiaojun Huang, Zhengfan Jiang
ZUSAMMENFASSUNG
Manganese (Mn) is essential for many physiological processes, but its functions in innate immunity remain undefined. Here, we found that Mn2+ was required for the host defense against DNA viruses by increasing the sensitivity of the DNA sensor cGAS and its downstream adaptor protein STING. Mn2+ was released from membrane-enclosed organelles upon viral infection and accumulated in the cytosol where it bound directly to cGAS. Mn2+ enhanced the sensitivity of cGAS to double-stranded DNA (dsDNA) and its enzymatic activity, enabling cGAS to produce secondary messenger cGAMP in the presence of low concentrations of dsDNA that would otherwise be non-stimulatory. Mn2+ also enhanced STING activity by augmenting cGAMP-STING binding affinity. Mn-deficient mice showed diminished cytokine production and were more vulnerable to DNA viruses, and Mn-deficient STING-deficient mice showed no increased susceptibility. These findings indicate that Mn is critically involved and required for the host defense against DNA viruses.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Histopaque®-1077, sterile-filtered, density: 1.077
Sigma-Aldrich
Monoklonaler ANTI-FLAG® M2-Antikörper in Maus hergestellte Antikörper, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
N-Acetyl-L-Cystein, Sigma Grade, ≥99% (TLC), powder
Sigma-Aldrich
α-Linoleic acid, ≥98%
Sigma-Aldrich
Monoklonaler ANTI-HA-Antikörper in Maus hergestellte Antikörper, clone HA-7, ascites fluid
Sigma-Aldrich
Pyridoxin, ≥98%
Sigma-Aldrich
Sera human, frozen liquid
Sigma-Aldrich
Acridinorange -hydrochlorid -Lösung, 10 mg/mL in H2O, ≥95.0% (HPLC)
Sigma-Aldrich
Maltose -Lösung, BioReagent, ~20% in H2O, Molecular Biology
Sigma-Aldrich
cGAMP sodium salt, ≥98% (HPLC)
Sigma-Aldrich
Anti-MB21D1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution