Merck
  • "Bottom-Up" Fabrication of BODIPY-Functionalized Fluorescent Hyperbranched Glycopolymers for Hepatoma-Targeted Imaging.
"Bottom-Up" Fabrication of BODIPY-Functionalized Fluorescent Hyperbranched Glycopolymers for Hepatoma-Targeted Imaging.
Macromolecular bioscience (2018-03-31)
Pei Sun, Hongping Deng, Linzhu Zhou, Yan Wu, Xin Jin, Gangsheng Tong, Xuemei Yu
ZUSAMMENFASSUNG
A novel type of multivalent and highly specific fluorescent hyperbranched glycopolymers h-P(GalEA-co-VBPT-co-BYMA) (hPGVB) is designed and prepared successfully via a facile "bottom-up" strategy. The acetylated hPGVB is prepared by one-pot reversible addition-fragmentation chain transfer (RAFT) copolymerization of acrylate-type galactose monomers AcGalEA and methacrylate-type fluorescent monomers BYMA in presence of an inimer-type RAFT chain transfer agent. After deacetylation, the resulting amphiphilic hPGVB can self-assemble into stable nanoparticles in aqueous media, showing strong green fluorescence with relative high quantum yields and good photostability. The cell viability study indicates the excellent biocompatibility of the hPGVB fluorescent nanoparticles (FNPs) against HepG2 and NIH3T3 cells. More importantly, comparing with the galactose-free fluorescent hyperbranched polymers h-P(OEGMA-co-VBPT-co-BYMA), hPEVB FNPs can be selectively internalized by asialoglycoprotein (ASGP) receptor-rich HepG2 cells, indicating their potential application in the bioimaging fields.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
2-Hydroxyethylacrylat, 96%, contains 200-650 ppm monomethyl ether hydroquinone as inhibitor
Sigma-Aldrich
2,4-Dimethylpyrrol, 97%