Merck
  • Quantitative structure-activity relationship study of phloroglucinol-terpene adducts as anti-leishmanial agents.
Quantitative structure-activity relationship study of phloroglucinol-terpene adducts as anti-leishmanial agents.
Bioorganic & Medicinal Chemistry Letters (2011)
Bharate SB, Singh IP.