Merck
  • Effects of corresponding and non-corresponding contaminants on the fate of sulfonamide and quinolone resistance genes in the Laizhou Bay, China.
Effects of corresponding and non-corresponding contaminants on the fate of sulfonamide and quinolone resistance genes in the Laizhou Bay, China.
Marine pollution bulletin (2018-03-25)
Qianwei Li, Guangshui Na, Linxiao Zhang, Zihao Lu, Hui Gao, Ruijing Li, Shuaichen Jin
ZUSAMMENFASSUNG
The environmental behaviors and migration patterns of antibiotic resistance genes (ARGs) have attracted considerable research interest. However, there has been little research concerning the effects of corresponding and non-corresponding contaminants on the fate of ARGs in coastal environments. In the present study, the distribution of intI1, sul1, sul2, qnrS and aac(6')-Ib were analyzed in water and sediment samples of Laizhou Bay in the context of corresponding and non-corresponding contaminants. The abundance of intI1, sul1 and sul2 genes exhibited a clear decreasing trend extending from the inner estuary to the coastal area. Strong and positive correlations existed between sul1/sul2 and sulfonamide antibiotic residues in sediments, and between the abundances of intI1 and sul1/sul2. Statistical analyses indicated that non-corresponding contaminants were partially correlated with ARG abundances. These results suggest that non-corresponding contaminants may have direct or indirect influences on the abundances of ARGs and intI1 in the Laizhou Bay.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
N-Octanoyl-N-Methylglucamin, ≥97% (GC)