Merck
  • Microwave-Assisted ZrSiO2 Catalysed Synthesis, Characterization and Computational Study of Novel Spiro[Indole-Thiazolidines] Derivatives as Anti-tubercular Agents.
Microwave-Assisted ZrSiO2 Catalysed Synthesis, Characterization and Computational Study of Novel Spiro[Indole-Thiazolidines] Derivatives as Anti-tubercular Agents.
Interdisciplinary sciences, computational life sciences (2016-11-12)
Mayuri A Borad, Manoj N Bhoi, Sanjay K Rathwa, Mahesh S Vasava, Hitesh D Patel, Chirag N Patel, Himanshu A Pandya, Edwin A Pithawala, John J Georrge
ZUSAMMENFASSUNG
In the current investigation, we prepared a series of novel spiro[indole-thiazolidines] derivatives (5a-5h) from 5-substituted isatin derivatives and thioglycolic acid (TGA) with ZrSiO2 as an efficient catalyst under microwave irradiation. The significant merits of this protocol have some significant merits such as simplicity in operation, simple, efficient workup, good practical yields of product and the employment of recyclable catalyst. All the new synthesized scaffold has been well characterized by various spectroscopic methods and elemental analysis. All the spiro scaffolds were subjected to in vitro anti-mycobacterial activity against the Mycobacterium tuberculosis (H37Rv) strain. We have carried out molecular docking study of our synthesized compounds. We also calculated theoretically ADME-Tox parameters for synthesized compounds.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
1,3-Propylene sulfite, 99%