Merck
  • Antioxidant and Neuroprotective Effects of N-((3,4-Dihydro-2H-benzo[h]chromen-2-yl)methyl)-4-methoxyaniline in Primary Cultured Rat Cortical Cells: Involvement of ERK-CREB Signaling.
Antioxidant and Neuroprotective Effects of N-((3,4-Dihydro-2H-benzo[h]chromen-2-yl)methyl)-4-methoxyaniline in Primary Cultured Rat Cortical Cells: Involvement of ERK-CREB Signaling.
Molecules (Basel, Switzerland) (2018-03-16)
Kyeongjun Lee, Chowee Park, Yeonsoo Oh, Heesoon Lee, Jungsook Cho
ZUSAMMENFASSUNG
Excitotoxicity and oxidative stress play vital roles in the development of neurodegenerative disorders including Alzheimer's disease (AD). In the present study, we investigated the effect of N-((3,4-dihydro-2H-benzo[h]chromen-2-yl)methyl)-4-methoxyaniline (BL-M) on excitotoxic neuronal cell damage in primary cultured rat cortical cells, and compared to that of memantine, a non-competitive N-methyl-d-aspartate (NMDA) receptor antagonist clinically used to treat AD. We found that BL-M inhibited glutamate- or N-methyl-d-aspartate (NMDA)-induced excitotoxic cell damage. The IC50 value of BL-M against NMDA toxicity was comparable to that of memantine. BL-M potently inhibited intracellular reactive oxygen species generated by glutamate or NMDA. Additionally, it inhibited the formation of 1,1-diphenyl-2-picryl-hydrazyl radicals in vitro and lipid peroxidation in rat brain homogenates. In contrast, memantine showed minimal or negligible antioxidant activity. Western blotting and immunocytochemical analyses showed that BL-M, not memantine, increased the ERK1/2 phosphorylation and subsequent phosphorylation of cAMP response element-binding protein (CREB). The inhibition of NMDA toxicity by BL-M was dramatically reversed by U0126, a well-known MEK inhibitor, suggesting that ERK1/2-mediated CREB phosphorylation is required for the neuroprotective action. Collectively, in this study, we demonstrated the neuroprotective effect of a newly synthesized chromene derivative BL-M and its underlying action mechanism(s). In contrast to memantine, BL-M exhibited marked antioxidant activity. Furthermore, it enhanced the ERK-mediated phosphorylation of CREB, which plays a crucial neuroprotective role. Our findings suggest that BL-M may be beneficial for AD and other neurodegenerative disorders associated with excitotoxicity as well as oxidative stress.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
N-Methyl-D-Asparaginsäure, ≥98% (TLC), solid