Merck
  • Binding of ferredoxin to algal photosystem I involves a single binding site and is composed of two thermodynamically distinct events.
Binding of ferredoxin to algal photosystem I involves a single binding site and is composed of two thermodynamically distinct events.
Biochimica et biophysica acta (2018-01-18)
Pini Marco, Marina Kozuleva, Haviva Eilenberg, Yuval Mazor, Peter Gimeson, Andrey Kanygin, Kevin Redding, Iddo Weiner, Iftach Yacoby
ZUSAMMENFASSUNG
Despite the impressive progress made in recent years in understanding the early steps in charge separation within the photosynthetic reaction centers, our knowledge of how ferredoxin (Fd) interacts with the acceptor side of photosystem I (PSI) is not as well developed. Fd accepts electrons after transiently docking to a binding site on the acceptor side of PSI. However, the exact location, as well as the stoichiometry, of this binding have been a matter of debate for more than two decades. Here, using Isothermal Titration Calorimetry (ITC) and purified components from wild type and mutant strains of the green algae Chlamydomonas reinhardtii we show that PSI has a single binding site for Fd, and that the association consists of two distinct binding events, each with a specific association constant.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Maltose -Lösung, BioReagent, ~20% in H2O, Molecular Biology