Merck
  • A Potent Anti-influenza Compound Blocks Fusion through Stabilization of the Prefusion Conformation of the Hemagglutinin Protein.
A Potent Anti-influenza Compound Blocks Fusion through Stabilization of the Prefusion Conformation of the Hemagglutinin Protein.
ACS infectious diseases (2015-05-20)
Kris M White, Paul De Jesus, Zhong Chen, Pablo Abreu, Elisa Barile, Puiying A Mak, Paul Anderson, Quy T Nguyen, Atsushi Inoue, Silke Stertz, Renate Koenig, Maurizio Pellecchia, Peter Palese, Kelli Kuhen, Adolfo García-Sastre, Sumit K Chanda, Megan L Shaw
ZUSAMMENFASSUNG
An ultrahigh-throughput screen was performed to identify novel small molecule inhibitors of influenza virus replication. The screen employed a recombinant influenza A/WSN/33 virus expressing Renilla luciferase and yielded a hit rate of 0.5%, of which the vast majority showed little cytotoxicity at the inhibitory concentration. One of the top hits from this screen, designated S20, inhibits HA-mediated membrane fusion. S20 shows potent antiviral activity (IC50 = 80 nM) and low toxicity (CC50 = 40 μM), yielding a selectivity index of 500 and functionality against all of the group 1 influenza A viruses tested in this study, including the pandemic H1N1 and avian H5N1 viruses. Mechanism of action studies proved a direct S20-HA interaction and showed that S20 inhibits fusion by stabilizing the prefusion conformation of HA. In silico docking studies were performed, and the predicted binding site in HA2 corresponds with the area where resistance mutations occurred and correlates with the known role of this region in fusion. This high-throughput screen has yielded many promising new lead compounds, including S20, which will potentially shed light on the molecular mechanisms of viral infection and serve as research tools or be developed for clinical use as antivirals.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
MBX2546, ≥98% (HPLC)