- A facile in-situ diazo-coupling reactions under two-phase conditions catalyzed by tetrakis [3, 5-bis (trifluoromethyl) phenyl] borate ion.
A facile in-situ diazo-coupling reactions under two-phase conditions catalyzed by tetrakis [3, 5-bis (trifluoromethyl) phenyl] borate ion.
Tetrahedron Letters (1983)
Iwamoto H, et al.