Merck
  • Injectable drug?delivery systems based on supramolecular hydrogels formed by poly (ethylene oxide) s and α?cyclodextrin.
Injectable drug?delivery systems based on supramolecular hydrogels formed by poly (ethylene oxide) s and α?cyclodextrin.
Journal of Biomedical Materials Research Part A (2003)
Li J N X and Leong K W