- Ceric-sulfate-catalyzed synthesis of 14-aryl-or 14-alkyl-14 H-dibenzo [aj] xanthene under conventional heating and microwave irradiation.
Ceric-sulfate-catalyzed synthesis of 14-aryl-or 14-alkyl-14 H-dibenzo [aj] xanthene under conventional heating and microwave irradiation.
Canadian Journal of Chemistry (2007)
Selvam N P, et al.