Merck
Low reliability of anti-KIR4.183-120 peptide auto-antibodies in multiple sclerosis patients.
Multiple sclerosis (Houndmills, Basingstoke, England) (2017-05-27)
Mariapaola Marino, Giovanni Frisullo, Gabriele Di Sante, Daniela Maria Samengo, Carlo Provenzano, Massimiliano Mirabella, Giovambattista Pani, Francesco Ria, Emanuela Bartoccioni
ZUSAMMENFASSUNG
Multiple sclerosis (MS) is an autoimmune disease for which auto-antibodies fully validated as diagnostic and prognostic biomarkers are widely desired. Recently, an immunoreactivity against the inward rectifying potassium channel 4.1 (KIR4.1) has been reported in a large proportion of a group of MS patients, with amino acids 83-120 being the major epitope. Moreover, a strong correlation between anti-KIR4.183-120 and anti-full-length-protein auto-antibodies titer was reported. However, this finding received limited confirmation. Validation of the diagnostic potential of anti-KIR4.183-120 antibodies in 78 MS patients, 64 healthy blood donors, and 42 individuals with other neurological diseases. Analysis of anti-KIR4.183-120 antibodies by enzyme-linked immunosorbent assay (ELISA) using a mouse antiserum we produced as a new ELISA reliability control. Additionally, evaluation of reactivity against 293-T cells transiently transfected with full-length KIR4.1 by flow cytometry. We found antibodies to KIR4.183-120 only in 13 out of 78 (16.6%) MS patients; among these, only 2 were positive for anti-full-length KIR4.1 antibodies. Employing a new reliability control and a new cytofluorometric assay, we cannot support anti-KIR4.183-120 auto-antibodies as a reliable biomarker in MS.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
L-(−)-Glukose, ≥99%