- Pre-early adenovirus 5 gene product regulates synthesis of early viral messenger RNAs.
Pre-early adenovirus 5 gene product regulates synthesis of early viral messenger RNAs.
Cell (1979-08-01)
A J Berk, F Lee, T Harrison, J Williams, P A Sharp
PMID487437