Merck
Fos-associated protein p39 is the product of the jun proto-oncogene.
Science (New York, N.Y.) (1988-05-20)
F J Rauscher, D R Cohen, T Curran, T J Bos, P K Vogt, D Bohmann, R Tjian, B R Franza
ZUSAMMENFASSUNG
The Fos protein complex and several Fos-related antigens (FRA) bind specifically to a sequence element referred to as the HeLa cell activator protein 1 (AP-1) binding site. A combination of structural and immunological comparisons has identified the Fos-associated protein (p39) as the protein product of the jun proto-oncogene (c-Jun). The p39/Jun protein is one of the major polypeptides identified in AP-1 oligonucleotide affinity chromatography extracts of cellular proteins. These preparations of AP-1 also contain Fos and several FRA's. Some of these proteins bind to the AP-1 site directly whereas others, like Fos, appear to bind indirectly via protein-protein interactions. Cell-surface stimulation results in an increase in c-fos and c-jun products. Thus, the products of two protooncogenes (and several related proteins), induced by extracellular stimuli, form a complex that associates with transcriptional control elements containing AP-1 sites, thereby potentially mediating the long-term responses to signals that regulate growth control and development.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
C-fos, proto oncogene human, recombinant, expressed in insect cells, ≥70% (SDS-PAGE)
Sigma-Aldrich
C-jun, proto oncogene human, recombinant, expressed in insect cells, ≥80% (SDS-PAGE)