Merck
  • Neuroprotective effects of 20(S)-protopanaxatriol (PPT) on scopolamine-induced cognitive deficits in mice.
Neuroprotective effects of 20(S)-protopanaxatriol (PPT) on scopolamine-induced cognitive deficits in mice.
Phytotherapy research : PTR (2018-02-23)
Cong Lu, Jingwei Lv, Liming Dong, Ning Jiang, Yan Wang, Qiong Wang, Yinghui Li, Shanguang Chen, Bei Fan, Fengzhong Wang, Xinmin Liu
ZUSAMMENFASSUNG
20(S)-protopanaxatriol (PPT), one of the ginsenosides from Panax ginseng, has been reported to have neuroprotective effects and to improve memory. The present study was designed to investigate the protective effect of PPT on scopolamine-induced cognitive deficits in mice. Male Institute of Cancer Research mice were pretreated with 2 different doses of PPT (20 and 40 μmol/kg) for 27 days by intraperitoneal injection, and scopolamine (0.75 mg/kg) was injected intraperitoneally for 9 days to induce memory impairment. Thirty minutes after the last pretreatment, the locomotor activity was firstly examined to evaluate the motor function of mice. Then, memory-related behaviors were evaluated, and the related mechanism was further researched. It was founded that PPT treatment significantly reversed scopolamine-induced cognitive impairment in the object location recognition experiment, the Morris water maze test, and the passive avoidance task, showing memory-improving effects. PPT also significantly improved cholinergic system reactivity and suppressed oxidative stress, indicated by inhibition of acetylcholinesterase activity, elevation of acetylcholine levels, increasing superoxide dismutase activity and lowering levels of malondialdehyde in the hippocampus. In addition, the expression levels of Egr-1, c-Jun, and cAMP responsive element binding in the hippocampus were significantly elevated by PPT administration. These results suggest that PPT may be a potential drug candidate for the treatment of cognitive deficit in Alzheimer's disease.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
CelLytic MT Zelllysereagens, For mammalian tissues