Merck
Polymer Nanosheet Containing Star-Like Copolymers: A Novel Scalable Controlled Release System.
Small (Weinheim an der Bergstrasse, Germany) (2018-04-28)
Peng-Fei Cao, Al de Leon, Lihan Rong, Ke-Zhen Yin, Eric C Abenojar, Zhe Su, Brylee David B Tiu, Agata A Exner, Eric Baer, Rigoberto C Advincula
ZUSAMMENFASSUNG
Poly(ε-caprolactone) (PCL)-based nanomaterials, such as nanoparticles and liposomes, have exhibited great potential as controlled release systems, but the difficulties in large-scale fabrication limit their practical applications. Among the various methods being developed to fabricate polymer nanosheets (PNSs) for different applications, such as Langmuir-Blodgett technique and layer-by-layer assembly, are very effort consuming, and only a few PNSs can be obtained. In this paper, poly(ε-caprolactone)-based PNSs with adjustable thickness are obtained in large quantity by simple water exposure of multilayer polymer films, which are fabricated via a layer multiplying coextrusion method. The PNS is also demonstrated as a novel controlled guest release system, in which release kinetics are adjustable by the nanosheet thickness, pH values of the media, and the presence of protecting layers. Theoretical simulations, including Korsmeyer-Peppas model and Finite-element analysis, are also employed to discern the observed guest-release mechanisms.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Zinn(II)-2-Ethylhexanoat, 92.5-100.0%