Merck
Sandwich-like Nanosystem for Simultaneous Removal of Cr(VI) and Cd(II) from Water and Soil.
ACS applied materials & interfaces (2018-05-08)
Dongfang Wang, Guilong Zhang, Zhangyu Dai, Linglin Zhou, Po Bian, Kang Zheng, Zhengyan Wu, Dongqing Cai
ZUSAMMENFASSUNG
In this work, a novel nanosystem with a sandwich-like structure was synthesized via face-to-face combination of two pieces of waste cotton fabrics (CFs) carrying ferrous sulfide (FeS) and carboxyl-functionalized ferroferric oxide microsphere (CFFM), respectively, and the obtained nanosystem was named as FeS/CFFM/CF. Therein, FeS has high reduction and adsorption capabilities for hexavalent chromium (Cr(VI)), CFFM possesses a high adsorption ability on cadmium ion (Cd(II)) through electrostatic attraction and chelation, and CF displays high immobilization ability for FeS and CFFM and adsorption performance on Cd(II). FeS/CFFM/CF could simultaneously remove Cr(VI) and Cd(II) from water and inhibit the uptake of Cr and Cd by fish and water spinach, ensuring the food safety. Besides, this technology could efficiently control the migration of Cr(VI) and Cd(II) in the sand-soil mixture, which was favorable to prevent their wide diffusion. Importantly, FeS/CFFM/CF possessed a high flexibility and could be conveniently produced with needed scale and shape and easily separated from water and soil, displaying a promising approach to remediate Cr(VI)-/Cd(II)-contaminated water and soil and a huge application potential.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natriumacrylat, 97%