Merck
  • Phytochemical and biotechnological studies on Schisandra chinensis cultivar Sadova No. 1-a high utility medicinal plant.
Phytochemical and biotechnological studies on Schisandra chinensis cultivar Sadova No. 1-a high utility medicinal plant.
Applied microbiology and biotechnology (2018-04-25)
Agnieszka Szopa, Marta Klimek-Szczykutowicz, Adam Kokotkiewicz, Anna Maślanka, Agata Król, Maria Luczkiewicz, Halina Ekiert
ZUSAMMENFASSUNG
In the presented work, raw materials (fruits and leaves) and in vitro biomass of a highly productive Schisandra chinensis Sadova No. 1 cultivar (SchS) were evaluated for the production of therapeutically useful schisandra lignans (SL). In vitro cultures of SchS were initiated, followed by extensive optimization studies focused on maximizing secondary metabolite production, with the aim of establishing a sustainable source of SL. Different cultivation systems (agar, agitated, bioreactor), experiment times (10, 20, 30, 40, 50 and 60 days) and plant growth regulators (6-benzyladenine-BA and 1-naphthaleneacetic acid-NAA, from 0 to 3 mg/l) in Murashige-Skoog (MS) medium were tested. Moreover, an elicitation procedure was applied to bioreactor-grown microshoots in order to increase SL production. Validated HPLC-DAD protocol enabled to detect fourteen SL in the extracts from in vitro and in vivo materials. The main compounds in the in vitro cultures were as follows: schisandrin (max. 176.3 mg/100 g DW), angeloylgomisin Q (max. 85.1 mg/100 g DW), gomisin A (max. 71.4 mg/100 g DW) and angeloylgomisin H (max. 67.0 mg/100 g DW). The highest total SL content (490.3 mg/100 g DW) was obtained in extracts from the biomass of agar cultures cultivated for 30 days on the MS medium variant containing 3 mg/l BA and 1 mg/l NAA. This amount was 1.32 times lower than in fruit extracts (646.0 mg/100 g DW) and 2.04 times higher than in leaf extracts (240.7 mg/100 g DW). The study demonstrated that SchS is a rich source of SL, thus proving its value for medical, cosmetic and food industry.