Merck
  • Purification and Characterization of Recombinant Botulinum Neurotoxin Serotype FA, Also Known as Serotype H.
Purification and Characterization of Recombinant Botulinum Neurotoxin Serotype FA, Also Known as Serotype H.
Toxins (2018-05-13)
Gavin Hackett, Kevin Moore, David Burgin, Fraser Hornby, Bryony Gray, Mark Elliott, Imran Mir, Matthew Beard
ZUSAMMENFASSUNG
We have purified and characterized recombinant botulinum neurotoxin serotype FA (BoNT/FA). This protein has also been named as a new serotype (serotype H), but the classification has been controversial. A lack of well-characterized, highly pure material has been a roadblock to study. Here we report purification and characterization of enzymatically active, and of inactive nontoxic, recombinant forms of BoNT/FA as tractable alternatives to purifying this neurotoxin from native Clostridium botulinum. BoNT/FA cleaves the same intracellular target proteins as BoNT/F1 and other F serotype BoNTs; the intracellular targets are vesicle associated membrane proteins (VAMP) 1, 2 and 3. BoNT/FA cleaves the same site in VAMP-2 as BoNT/F5, which is different from the cleavage site of other F serotype BoNTs. BoNT/FA has slower enzyme kinetics than BoNT/F1 in a cell-free protease assay and is less potent at inhibiting ex vivo nerve-stimulated skeletal muscle contraction. In contrast, BoNT/FA is more potent at inhibiting neurotransmitter release from cultured neurons.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-SNAP-25 in Kaninchen hergestellte Antikörper, IgG fraction of antiserum, buffered aqueous solution