Merck
  • The vanillin derivative 6-bromine-5-hydroxy-4-methoxybenzaldehyde induces aberrant mitotic progression and enhances radio-sensitivity accompanying suppression the expression of PLK1 in esophageal squamous cell carcinoma.
The vanillin derivative 6-bromine-5-hydroxy-4-methoxybenzaldehyde induces aberrant mitotic progression and enhances radio-sensitivity accompanying suppression the expression of PLK1 in esophageal squamous cell carcinoma.
Toxicology and applied pharmacology (2018-04-22)
Meng-Meng Gu, Ming Li, Dexuan Gao, Lang-Huan Liu, Yue Lang, Si-Ming Yang, Hongling Ou, Bo Huang, Ping-Kun Zhou, Zeng-Fu Shang
ZUSAMMENFASSUNG
Esophageal squamous cell carcinoma (ESCC) is the most common form of esophageal cancer in China. Since chemotherapy is the standard clinical intervention for advanced ESCC, the development of highly effective and minimal/non-toxic drugs is essential to improve the clinical outcome and prognosis of the patients. A novel derivative of vanillin, 6-bromine-5-hydroxy-4-methoxybenzaldehyde (BVAN08), has been recently reported to activate different cell death pathways in cancer cells. In this study, we demonstrate that BVAN08 exhibits a potent anti-proliferation effect on ESCC cells (TE-1 and ECA-109) by inhibiting the expression of PLK1, an important mitotic kinase. Consistent with this, BVAN08 induces mitotic arrest and chromosomal misalignment in ESCC cells. The disruption of microtubule nucleation around centrosomes is also observed in BVAN08 treated ESCC cells. Furthermore, BVAN08 enhances radio-sensitivity of ESCC cells by prolonging DNA damage repair. These findings underscore the potential value of BVAN08 in cancer therapeutics and demonstrate the underlying mechanism by which BVAN08 induces mitotic catastrophe and enhances radio-sensitivity in ESCC cells.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Nocodazol, ≥99% (TLC), powder