Merck
  • Photophysics of Thermally-Assisted Photobleaching in "Giant" Quantum Dots Revealed in Single Nanocrystals.
Photophysics of Thermally-Assisted Photobleaching in "Giant" Quantum Dots Revealed in Single Nanocrystals.
ACS nano (2018-05-02)
Noah J Orfield, Somak Majumder, James R McBride, Faith Yik-Ching Koh, Ajay Singh, Sarah J Bouquin, Joanna L Casson, Alex D Johnson, Liuyang Sun, Xiaoqin Li, Chih-Kang Shih, Sandra J Rosenthal, Jennifer A Hollingsworth, Han Htoon
ZUSAMMENFASSUNG
Quantum dots (QDs) are steadily being implemented as down-conversion phosphors in market-ready display products to enhance color rendering, brightness, and energy efficiency. However, for adequate longevity, QDs must be encased in a protective barrier that separates them from ambient oxygen and humidity, and device architectures are designed to avoid significant heating of the QDs as well as direct contact between the QDs and the excitation source. In order to increase the utility of QDs in display technologies and to extend their usefulness to more demanding applications as, for example, alternative phosphors for solid-state lighting (SSL), QDs must retain their photoluminescence emission properties over extended periods of time under conditions of high temperature and high light flux. Doing so would simplify the fabrication costs for QD display technologies and enable QDs to be used as down-conversion materials in light-emitting diodes for SSL, where direct-on-chip configurations expose the emitters to temperatures approaching 100 °C and to photon fluxes from 0.1 W/mm2 to potentially 10 W/mm2. Here, we investigate the photobleaching processes of single QDs exposed to controlled temperature and photon flux. In particular, we investigate two types of room-temperature-stable core/thick-shell QDs, known as "giant" QDs for which shell growth is conducted using either a standard layer-by-layer technique or by a continuous injection method. We determine the mechanistic pathways responsible for thermally-assisted photodegradation, distinguishing effects of hot-carrier trapping and QD charging. The findings presented here will assist in the further development of advanced QD heterostructures for maximum device lifetime stability.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Trioctylphosphin, 97%
Sigma-Aldrich
Trioctylphosphinoxid, ReagentPlus®, 99%
Sigma-Aldrich
1-Octanthiol, ≥98.5%
Sigma-Aldrich
Octadecylphosphonsäure, 97%
Sigma-Aldrich
Cadmiumoxid, ≥99.99% trace metals basis