Merck
  • Mechanical and spectroscopic characterization of crosslinked zein films cast from solutions of acetic acid leading to a new mechanism for the crosslinking of oleic acid plasticized zein films.
Mechanical and spectroscopic characterization of crosslinked zein films cast from solutions of acetic acid leading to a new mechanism for the crosslinking of oleic acid plasticized zein films.
Food research international (Ottawa, Ont.) (2018-05-08)
Hazal Turasan, Emma A Barber, Morgan Malm, Jozef L Kokini
ZUSAMMENFASSUNG
This study discovered through FTIR, FT-Raman and rheological measurements that glutaraldehyde binds to zein through the amine groups of glutamine turns by replacing the already-bonded oleic acid molecules and forming imine structures. As a secondary crosslinking mechanism, glutaraldehyde unfolds some of the α-helices and turns them into β-sheets. While crosslinking resulted in stiffer and less ductile zein films, it made the surface of the films rougher, measured using AFM, and more hydrophilic, measured using WCA. In the crosslinking conditions in this study, the number of crosslinks estimated from rubber elasticity theory were not enough to change the water vapor permeability of the films significantly. Improving the understanding of crosslinking mechanism and its effects on physical and chemical properties of zein films can be useful to develop stiffer, stronger and more durable platforms for biodegradable biosensors, microfluidic devices or scaffolds.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Oleinsäure, technical grade, 90%