Merck
  • Inhibiting aerobic glycolysis suppresses renal interstitial fibroblast activation and renal fibrosis
Inhibiting aerobic glycolysis suppresses renal interstitial fibroblast activation and renal fibrosis
American Journal of Physiology: Renal Physiology (2017)
Ding H, et al.