Merck
  • Synthesis of Lanostane-Type Triterpenoid N-Glycosides and Their Cytotoxicity against Human Cancer Cell Lines.
Synthesis of Lanostane-Type Triterpenoid N-Glycosides and Their Cytotoxicity against Human Cancer Cell Lines.
Chemistry & biodiversity (2018-05-08)
Motohiko Ukiya, Teppei Hayakawa, Kouta Okazaki, Maiko Hikawa, Hiroyuki Akazawa, Wei Li, Kazuo Koike, Makoto Fukatsu
ZUSAMMENFASSUNG
Seventeen lanostane-type triterpenoid derivatives (2 - 18), including 11N-glycosides (8 - 18), were synthesized from the natural triterpenoid, lanosterol (1), and were evaluated for their cytotoxicity against the human cancer cell lines, HL-60, A549, and MKN45, as well as the normal human lung cells, WI-38. Among them, N-β-d-2-acetamido-2-deoxyglucoside (10) showed cytotoxicity against HL-60, A549, MKN45, and WI-38 cells (IC50 0.0078 - 2.8 μm). However, N-β-d-galactoside (12) showed cytotoxicity against HL-60 and MKN45 cells (IC50 0.0021 - 4.0 μm), but not the normal WI-38 cells. Furthermore, Western blot analysis suggested that 12 induces apoptosis by activation of caspases-3, 8, and 9. These results will be useful for the synthesis of other tetracyclic triterpenoids or steroid N-glycosides to increase their cytotoxicity and apoptosis-inducing activities.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
L-(−)-Glukose, ≥99%
Sigma-Aldrich
D-(+)-Fucose, ≥97% (GC)
Sigma-Aldrich
Lanosterin, ≥93%, powder
Sigma-Aldrich
D-(+)-Talose, ≥99.0% (HPLC)