Merck
  • Thionations using a P4S10-pyridine complex in solvents such as acetonitrile and dimethyl sulfone.
Thionations using a P4S10-pyridine complex in solvents such as acetonitrile and dimethyl sulfone.
The Journal of organic chemistry (2011-02-24)
Jan Bergman, Birgitta Pettersson, Vedran Hasimbegovic, Per H Svensson
ZUSAMMENFASSUNG
Tetraphosphorus decasulfide (P(4)S(10)) in pyridine has been used as a thionating agent for a long period of time. The moisture-sensitive reagent has now been isolated in crystalline form, and the detailed structure has been determined by X-ray crystallography. The thionating power of this storable reagent has been studied and transferred to solvents such as acetonitrile in which it has proven to be synthetically useful and exceptionally selective. Its properties have been compared with the so-called Lawesson reagent (LR). Particularly interesting are the results from thionations at relatively high temperatures (∼165 °C) in dimethyl sulfone as solvent. Under these conditions, for instance, acridone and 3-acetylindole could quickly be transformed to the corresponding thionated derivatives. Glycylglycine similarly gave piperazinedithione. At these temperatures, LR is inefficient due to rapid decomposition. The thionated products are generally cleaner and more easy to obtain because in the crystalline reagent, impurities which invariably are present in the conventional reagents, P(4)S(10) in pyridine or LR, have been removed.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dimethylsulfon, 98%
Supelco
Dimethylsulfon, qNMR standard, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland