Merck
  • Synthesis and Electrochemistry of O3-type NaFeO2-NaCo0.5Ni0.5O2 Solid Solutions for Na-Ion Positive Electrodes.
Synthesis and Electrochemistry of O3-type NaFeO2-NaCo0.5Ni0.5O2 Solid Solutions for Na-Ion Positive Electrodes.
ACS applied materials & interfaces (2018-06-14)
J S Thorne, Lituo Zheng, Christopher L D Lee, R A Dunlap, M N Obrovac
ZUSAMMENFASSUNG
The synthesis, structure, and electrochemistry in Na cells of NaFe xM1- xO2 positive electrode materials with M = Ni, Co0.5Ni0.5, and Co are reported. In particular, the properties of O3-NaFeO2-NaCo0.5Ni0.5O2 solid solutions having compositions NaFe x(Co0.5Ni0.5)1- xO2 with 0 ≤ x ≤ 0.5 are explored. It is found that the substitution of Fe in NaNi0.5Co0.5O2 causes an increase in first cycle energy density from 320 to 440 mWh/g in a 1.5-4.0 V test. However, capacity retention is generally reduced when x is increased for all M = Ni, Co0.5Ni0.5, and Co. In general, NaFe xM1- xO2 samples with M = Co had the highest capacity retention for all values of x. Ex situ X-ray diffraction and Mössbauer results of as-prepared and charged materials are directly compared for NaFe x(Co0.5Ni0.5)1- xO2 and NaFe xCo1- xO2 ( x = 0.4, 0.5). Iron was found to be in the +3 oxidation state in the as-prepared materials. A significant fraction of Fe3+ is oxidized to Fe4+ in these samples when they are charged to 4.0 V.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
1-Methyl-2-pyrrolidon, anhydrous, 99.5%
Sigma-Aldrich
Fluorethylencarbonat, 99%