Merck
  • Photophysicochemical properties and photodynamic therapy activity of highly water-soluble Zn(II) phthalocyanines.
Photophysicochemical properties and photodynamic therapy activity of highly water-soluble Zn(II) phthalocyanines.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2018-06-06)
David O Oluwole, Fatma Aslıhan Sarı, Earl Prinsloo, Edith Dube, Abdulcelil Yuzer, Tebello Nyokong, Mine Ince
ZUSAMMENFASSUNG
The syntheses of two zinc(II) phthalocyanines (ZnPcs) having either imidazole (ZnPc 1) or pyridiloxy (ZnPc 2) moieties as their macrocycle substituents are reported. Quaternization of the ZnPcs with methyl iodide afforded water soluble cationic phthalocyanines. The photophysical, photochemical properties and photodynamic therapy (PDT) activity of the ZnPcs were studied in solution. The fluorescence quantum yield and lifetime of ZnPc 1 were higher as compared to ZnPc 2. ZnPc 2 afforded higher triplet state (ΦT) and singlet oxygen quantum yields (ΦΔ) in comparison to ZnPc 1. The PDT activity of ZnPcs was investigated against human breast adenocarcinoma cells (MCF-7). The two compounds afforded a very minimal in vitro dark cytotoxicity with 85% viable cells at concentration ≤80 μM. On irradiation of the cells having the ZnPcs, ≥50% cell death was recorded for ZnPc 1 which was also evidenced by the cells photo-micrograph.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
1,3-Diphenyl-isobenzofuran, 97%