Merck
Biotransformation of cyclocanthogenol by the endophytic fungus Alternaria eureka 1E1BL1.
Phytochemistry (2018-04-21)
Güner Ekiz, Seda Duman, Erdal Bedir
ZUSAMMENFASSUNG
The microbial transformation of cyclocanthogenol (CCG), Astragalus sp. originated sapogenin, by the endophytic fungus Alternaria eureka 1E1BL1 isolated from Astragalus angustifolius was investigated. Hydroxylation, oxidation, epoxidation, O-methylation, ring-expansion and methyl migration reactions were observed on the triterpenoid skeleton. As a result, eight metabolites were isolated and the structures of the previously undescribed compounds were established by 1-D, 2-D NMR and HR-MS analyses.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
3-(Benzyldimethylammonio)-propansulfonat, BioXtra, ≥99.0% (HPCE)