Merck
  • In vitro evaluation of chitosan copper chelate gels as a multimicronutrient feed additive for cattle.
In vitro evaluation of chitosan copper chelate gels as a multimicronutrient feed additive for cattle.
Journal of the science of food and agriculture (2018-02-09)
Ciara Duffy, Dolores O'Riordan, Michael O'Sullivan, Jean-Christophe Jacquier
ZUSAMMENFASSUNG
Effective micronutrient supplementation strategies are critical to ensure optimal health and productivity in livestock. The objective of this study was to develop a copper and vitamin (multimicronutrient) delivery system based on chitosan gel beads, and test its suitability, in vitro, for use as a cattle feed additive. Chitosan was chelated with copper sulfate to produce millimetre-scale gel matrices (∼2 mm). The copper content was significantly increased (from 61 to 95 mg g by adjusting pH to alkaline conditions post bead formation. The beads could subsequently be loaded with the model vitamin riboflavin to levels as high as 324 µg g-1 beads. Restricted rehydration of the dried gel matrices in simulated rumen fluid led to a sustained release of riboflavin with no copper released in these neutral conditions for up to 24 h, demonstrating copper rumen bypass. Moreover, sustained release of the mineral was observed in abomasal conditions of pH 2 over a 3 h period. The matrices showed rumen bypass for copper yet supplied nutritionally relevant levels of the free mineral in abomasal conditions, as required for effective supplementation in cattle. The controlled-release properties demonstrated by the matrices indicate their potential as a multimicronutrient functional feed additive to enhance cattle nutrition and productivity. © 2018 Society of Chemical Industry.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
meso-Tetraphenylporphyrin, BioReagent, suitable for fluorescence, ≥99.0% (HPLC)