Merck
  • Debranching of pea starch using pullulanase and ultrasonication synergistically to enhance slowly digestible and resistant starch.
Debranching of pea starch using pullulanase and ultrasonication synergistically to enhance slowly digestible and resistant starch.
Food chemistry (2018-08-02)
Zhan-Hui Lu, Nicholas Belanger, Elizabeth Donner, Qiang Liu
ZUSAMMENFASSUNG
Pullulanase (P) and ultrasonication (U) were simultaneously applied to debranch pea starch to enhance slowly digestible starch (SDS) and resistant starch (RS) fractions in the debranched pea starch (DPS). A synergistic debranching effect was found under conditions of pullulanase (40 npun/g) and ultrasonication (100% amplitude in pulse mode, 1 min on followed by 9 min off), which produced 73.5% linear glucans, 18% SDS and 26% RS in the resulting DPS-PU after 6 h of debranching. Even when autoclaving the DPS-PU at 118 °C for 30 min, following cooldown, 11% SDS and 25% RS were retained in the DPS-PU, compared with 0% SDS and 12% RS in autoclaved native pea starch. The SDS fraction in autoclaved DPS-PU further increased to 16% while the RS content remained constant during 14 days of cold storage. In summary, DPS-PU is high in linear glucans, low in starch digestibility and has a thermally stable RS fraction.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Pancreatin aus Schweinepankreas, 8 × USP specifications
Sigma-Aldrich
Pepsin aus Schweinemagenschleimhaut, powder, ≥400 units/mg protein
Sigma-Aldrich
Invertase aus Backhefe (S. cerevisiae), Grade VII, ≥300 units/mg solid