Merck
  • Spectroscopic insight into supramolecular assemblies of boric acid derivatives and β-cyclodextrin.
Spectroscopic insight into supramolecular assemblies of boric acid derivatives and β-cyclodextrin.
Carbohydrate polymers (2018-08-11)
Artur Kasprzak, Krzysztof M Borys, Sergey Molchanov, Agnieszka Adamczyk-Woźniak
ZUSAMMENFASSUNG
Interactions of select boric acid derivatives with β-cyclodextrin were investigated. All products were obtained employing the grinding-induced mechanochemical approach. It was found that phenylboronic acid, benzoxaborole and boric acid form non-covalent, hydrogen bonding-based systems with β-cyclodextrin, whereas catechol and pinacol esters of phenylboronic acid as well as ferroceneboronic acid form host-guest inclusion complexes. The interactions were probed using spectroscopic methods: 1H NMR, 1H-1H ROESY NMR, 1H DOSY NMR, FT-IR. Association constant values were evaluated by 1H DOSY NMR. The highest association constant was found for boric acid (117.5 ± 2.5 M-1), whilst the lowest for benzoxaborole (14.8 ± 0.3 M-1). The study shows the influence of boron compound structure on the nature of the assembly formed with β-cyclodextrin, laying up the basis for future work with such supramolecular systems.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Phenylboronsäure, 95%