Merck
  • In vitro and in vivo effect of 2,6-Di-tert-butyl-4-methylphenol as an antibiofilm agent against quorum sensing mediated biofilm formation of Vibrio spp.
In vitro and in vivo effect of 2,6-Di-tert-butyl-4-methylphenol as an antibiofilm agent against quorum sensing mediated biofilm formation of Vibrio spp.
International journal of food microbiology (2018-06-05)
Sivasubramanian Santhakumari, Rengarajan Jayakumar, Ravichandran Logalakshmi, Narayanan Marimuthu Prabhu, Abdul Kuthus Abdul Nazar, Shunmugiah Karutha Pandian, Arumugam Veera Ravi
ZUSAMMENFASSUNG
This study unveils the in vitro and in vivo antibiofilm potential of 2,6-Di-tert-butyl-4-methylphenol (DTBMP) from Chroococcus turgidus against Vibrio spp. In the preliminary study, cell free culture supernatant (CFCS) of C. turgidus inhibited the violacein production in biomarker strain Chromobacterium violaceum and its mutant strain CV026 in a dose dependent manner. The effective biofilm inhibitory concentration (BIC) of pure compound DTBMP from C. turgidus was identified as 250 μg/ml concentration in tested Vibrio species. Furthermore, DTBMP proved to effectively inhibit the bioluminescence production in V. harveyi and other biofilm related virulence traits such as exopolysaccharides (EPS) production, hydrophobicity index, swimming and swarming motility at its BIC concentration in three major pathogenic vibrios: V. harveyi, V. parahaemolyticus and V. vulnificus. The antibiofilm potential of DTBMP was validated through light, confocal laser scanning and scanning electron microscopic analyses. In addition, the non-bactericidal effect of DTBMP was determined through growth curve and 2,3-bis (2-methyloxy-4-nitro-5-sulfophenyl)-2H-tetrazolium-5-carboxanilide (XTT) assay. Real-time PCR studies revealed the down-regulation of master quorum sensing (QS) regulator genes of V. harveyi such as luxR, luxS, luxP, luxQ and luxO on treatment with DTBMP. In vivo results confirmed that DTBMP augmented the survival rate of Litopenaeus vannamei larvae up to 75, 88 and 66% upon infection with V. harveyi, V. parahaemolyticus and V. vulnificus, respectively. The results of this study ascertain the promising effects of DTBMP as an antibiofilm agent, which could be positively explored to treat biofilm-associated vibrios infections in aquaculture.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
2,6-Di-tert.-butyl-4-methyl-pyridin, 98%
Sigma-Aldrich
N-Hexanoyl-L-homoserin-lacton, ≥96% (HPLC)