Merck
Isoform selectivity of harmine-conjugated 1,2,3-triazoles against human monoamine oxidase.
Future medicinal chemistry (2018-05-24)
Saqlain Haider, Manal Alhusban, Narayan D Chaurasiya, Babu L Tekwani, Amar G Chittiboyina, Ikhlas A Khan
ZUSAMMENFASSUNG
There is little information available on the monoamine oxidase isoform selectivity of N-alkyl harmine analogs, which exhibit a myriad of activities including MAO-A, DYRK1A and cytotoxicity to several select cancer cell lines. Compounds 3e and 4c exhibited an IC50 of 0.83 ± 0.03 and 0.43 ± 0.002 μM against MAO-A and an IC50 of 0.26 ± 0.04 and 0.36 ± 0.001 μM against MAO-B, respectively. Molecular docking studies revealed π-π interactions between the synthesized molecules and aromatic amino acid residues. Conclusion & future perspective: The current study delineates the structural requirements for MAO-A selectivity and such information may be helpful in designing selective analogs for kinase, DYRK1A and harmine-based cytotoxics without apparent MAO enzyme inhibition.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Harmin, 98%