Merck
Aspirin restores ABT-737-mediated apoptosis in human renal carcinoma cells.
Biochemical and biophysical research communications (2018-05-25)
Yen-Chuan Ou, Jian-Ri Li, Jiaan-Der Wang, Wen-Ying Chen, Yu-Hsiang Kuan, Ching-Ping Yang, Su-Lan Liao, Hsi-Chi Lu, Chun-Jung Chen
ZUSAMMENFASSUNG
Aspirin is a novel chemopreventive agent against malignancy. However, outcomes of aspirin monotherapy of renal cell carcinoma (RCC) are inconsistent across studies. ABT-737, an BH3 mimetic inhibitor, is also a promising antitumor drug. Cancer cells including those from RCC, that have high levels of Mcl-1, are refractory to ABT-737-induced apoptosis. We here investigated how aspirin treatment modulates the ABT-737-induced apoptosis. Using the in vitro model of human 786-O cells, we showed that aspirin had sensitized cells to ABT-737 induced apoptosis. Such aspirin-induced changes of ABT-737 resistance was accompanied by a host of biochemical events like protein phosphatase 2A (PP2A) activation, AKT dephosphorylation, Mcl-1/FLICE inhibiting protein (FLIP)/XIAP downregulation, and Bax mitochondrial redistribution. The PP2A inhibitor, okadaic acid, was able to reverse the apirin-induced apoptotic changes. Apart from the aspirin treatment, Mcl-1 silencing also rendered cells vulnerable to ABT-737 induced apoptosis. Since PP2A, Akt, and Mcl-1 play critical roles in RCC malignancy and treatment resistance, our present study showed that aspirin, an alternative adjuvant agent, had recalled ABT-737 sensitivity in the RCC cells through processes involving the PP2A/Akt/Mcl-1 axis.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
DL-Glyceraldehyd-3-phosphat -Lösung, 45-55 mg/mL in H2O