Merck
  • Moderate UV Exposure Enhances Learning and Memory by Promoting a Novel Glutamate Biosynthetic Pathway in the Brain.
Moderate UV Exposure Enhances Learning and Memory by Promoting a Novel Glutamate Biosynthetic Pathway in the Brain.
Cell (2018-05-22)
Hongying Zhu, Ning Wang, Lei Yao, Qi Chen, Ran Zhang, Junchao Qian, Yiwen Hou, Weiwei Guo, Sijia Fan, Siling Liu, Qiaoyun Zhao, Feng Du, Xin Zuo, Yujun Guo, Yan Xu, Jiali Li, Tian Xue, Kai Zhong, Xiaoyuan Song, Guangming Huang, Wei Xiong
ZUSAMMENFASSUNG
Sunlight exposure is known to affect mood, learning, and cognition. However, the molecular and cellular mechanisms remain elusive. Here, we show that moderate UV exposure elevated blood urocanic acid (UCA), which then crossed the blood-brain barrier. Single-cell mass spectrometry and isotopic labeling revealed a novel intra-neuronal metabolic pathway converting UCA to glutamate (GLU) after UV exposure. This UV-triggered GLU synthesis promoted its packaging into synaptic vesicles and its release at glutamatergic terminals in the motor cortex and hippocampus. Related behaviors, like rotarod learning and object recognition memory, were enhanced after UV exposure. All UV-induced metabolic, electrophysiological, and behavioral effects could be reproduced by the intravenous injection of UCA and diminished by the application of inhibitor or short hairpin RNA (shRNA) against urocanase, an enzyme critical for the conversion of UCA to GLU. These findings reveal a new GLU biosynthetic pathway, which could contribute to some of the sunlight-induced neurobehavioral changes.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-NeuN-Antikörper, Klon A60, clone A60, Chemicon®, from mouse
Sigma-Aldrich
Anti-Glyceraldehyd-3-phosphat-dehydrogenase-Antikörper, Klon 6C5, clone 6C5, Chemicon®, from mouse
Sigma-Aldrich
4-Imidazolacrylsäure, 99%
Sigma-Aldrich
Anti-Golgi FTCD antibody, clone 58k-9, Mouse monoclonal, ascites fluid
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Uroc1