Merck
  • Profile of Polyphenolic and Essential Oil Composition of Polish Propolis, Black Poplar and Aspens Buds.
Profile of Polyphenolic and Essential Oil Composition of Polish Propolis, Black Poplar and Aspens Buds.
Molecules (Basel, Switzerland) (2018-05-26)
Piotr Okińczyc, Antoni Szumny, Jakub Szperlik, Anna Kulma, Roman Franiczek, Beata Żbikowska, Barbara Krzyżanowska, Zbigniew Sroka
ZUSAMMENFASSUNG
In this work, we studied similarities and differences between 70% ethanol in water extract (70EE) and essential oils (EOs) obtained from propolis, black poplars (Populus nigra L.) and aspens (P. tremula L.) to ascertain which of these is a better indicator of the plant species used by bees to collect propolis precursors. Composition of 70EE was analyzed by UPLC-PDA-MS, while GC-MS was used to research the EOs. Principal component analyses (PCA) and calculations of Spearman's coefficient rank were used for statistical analysis. Statistical analysis exhibited correlation between chemical compositions of propolis and Populus buds' 70EE. In the case of EOs, results were less clear. Compositions of black poplars, aspens EOs and propolises have shown more variability than 70EE. Different factors such as higher instability of EOs compared to 70EE, different degradation pattern of benzyl esters to benzoic acid, differences in plant metabolism and bees' preferences may be responsible for these phenomena. Our research has therefore shown that 70EE of propolis reflected the composition of P. nigra or complex aspen⁻black poplar origin.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Pinobanksin, ≥95% (HPLC)