Merck
  • Arterial wall hypertrophy is ameliorated by α2-adrenergic receptor antagonist or aliskiren in kidneys of angiotensinogen-knockout mice.
Arterial wall hypertrophy is ameliorated by α2-adrenergic receptor antagonist or aliskiren in kidneys of angiotensinogen-knockout mice.
Clinical and experimental nephrology (2017-12-13)
Haruka Nakamori, Shin-Ichiro Yoshida, Hiroaki Ishiguro, Shota Suzuki, Hiroaki Yasuzaki, Tatsuo Hashimoto, Tomoaki Ishigami, Nobuhito Hirawa, Yoshiyuki Toya, Satoshi Umemura, Kouichi Tamura
ZUSAMMENFASSUNG
Arterial hypertrophy and interstitial fibrosis are important characteristics in kidneys of angiotensinogen-knockout (Atg -/-) mice. In these mice, which exhibit polyuria and hypotension, sympathetic nerve signaling is estimated to be compensatorily hyperactive. Furthermore, transforming growth factor (TGF)-β1 is overexpressed in mice kidneys. To determine whether sympathetic nerve signaling and TGF-β1 exacerbate arterial hypertrophy and interstitial fibrosis, intervention studies of such signaling are required. We performed renal denervation and administered the α2-adrenergic receptor (AR) antagonist, atipamezole, to Atg -/- mice. A renin inhibitor, aliskiren, which was preliminarily confirmed to reduce TGF-β1 gene expression in kidneys of the mice, was additionally administered to assess the effect on the arterial hypertrophy and interstitial fibrosis. Norepinephrine content in kidneys of Atg -/- mice was three times higher than in kidneys of wild-type mice. Interventions by renal denervation and atipamezole resulted in amelioration of the histological findings. Overexpression of TGF-β1 gene in kidneys of Atg -/- mice was altered in a manner linked to the histological findings. Surprisingly, aliskiren reduced α2-AR gene expression, interstitial fibrosis, and arterial hypertrophy in kidneys of Atg -/- mice, which lack renin substrate. Alpha2-AR signaling is one of the causes of persistent renal arterial hypertrophy in Atg -/- mice. Aliskiren also angiotensinogen-independently reduces the extent of renal arterial hypertrophy, partly thorough downregulation of α2-ARs. Although renal arterial hypertrophy in Atg -/- mice appears to be of multifactorial origin, TGF-β1 may play a key role in the persistence of such hypertrophy.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Aliskiren