Merck
New 'chemical probes' to examine the role of the hFPRL1 (or ALXR) receptor in inflammation.
Bioorganic & medicinal chemistry letters (2007-10-09)
Mike Frohn, Han Xu, Xiaoming Zou, Catherine Chang, Michele McElvaine, Matthew H Plant, Min Wong, Philip Tagari, Randall Hungate, Roland W Bürli
ZUSAMMENFASSUNG
We report the development of the novel N-substituted benzimidazole 11 as a potent and selective human formyl peptide receptor-like 1 (hFPRL1) agonist. This compound and its less active enantiomer 12 were identified as useful tools for studying receptor function in vitro.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
4-(2-Keto-1-benzimidazolinyl)piperidin, 98%