Merck
  • Discovery of a Selective Allosteric Inhibitor Targeting Macrodomain 2 of Polyadenosine-Diphosphate-Ribose Polymerase 14.
Discovery of a Selective Allosteric Inhibitor Targeting Macrodomain 2 of Polyadenosine-Diphosphate-Ribose Polymerase 14.
ACS chemical biology (2017-10-11)
Marion Schuller, Kerstin Riedel, Ian Gibbs-Seymour, Kristin Uth, Christian Sieg, André P Gehring, Ivan Ahel, Franz Bracher, Benedikt M Kessler, Jonathan M Elkins, Stefan Knapp
ZUSAMMENFASSUNG
Macrodomains are conserved protein interaction modules that can be found in all domains of life including in certain viruses. Macrodomains mediate recognition of sequence motifs harboring adenosine diphosphate ribose (ADPR) modifications, thereby regulating a variety of cellular processes. Due to their role in cancer or viral pathogenesis, macrodomains have emerged as potential therapeutic targets, but the unavailability of small molecule inhibitors has hampered target validation studies so far. Here, we describe an efficient screening strategy for identification of small molecule inhibitors that displace ADPR from macrodomains. We report the discovery and characterization of a macrodomain inhibitor, GeA-69, selectively targeting macrodomain 2 (MD2) of PARP14 with low micromolar affinity. Co-crystallization of a GeA-69 analogue with PARP14 MD2 revealed an allosteric binding mechanism explaining its selectivity over other human macrodomains. We show that GeA-69 engages PARP14 MD2 in intact cells and prevents its localization to sites of DNA damage.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
GeA-69, ≥98% (HPLC)