Merck
Hydroxamates: relationships between structure and plasma stability.
Journal of medicinal chemistry (2009-10-14)
Marion Flipo, Julie Charton, Akila Hocine, Sandrine Dassonneville, Benoit Deprez, Rebecca Deprez-Poulain
ZUSAMMENFASSUNG
Hydroxamates are valuable tools for chemical biology as well as interesting leads for medicinal chemistry. Although many hydroxamates display nanomolar activities against metalloproteases, only three hydroxamates have reached the market, among which is the HDAC inhibitor vorinostat. Failures in development are generally attributed to lack of selectivity, toxicity, or poor stability. To help medicinal chemists with respect to plasma stability, we have performed the first and preliminary study on structure-plasma stability for hydroxamates. We define some structural rules to predict or improve the plasma stability in the preclinical stage.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Benzhydroxamsäure, 99%