Merck
Controllable synthesis and tunable photocatalytic properties of Ti3+-doped TiO2
Scientific Reports (2015)
Ren, Ren and Wen, Zhenhai and Cui, Shumao and Hou, Yang and Guo, Xiaoru and Chen, Junhong