Merck
  • Development and optimization of a HS-SPME-GC-MS methodology to quantify volatile carbonyl compounds in Port wines.
Development and optimization of a HS-SPME-GC-MS methodology to quantify volatile carbonyl compounds in Port wines.
Food chemistry (2018-09-04)
Nathalie Moreira, Ana Margarida Araújo, Frank Rogerson, Isabel Vasconcelos, Victor De Freitas, Paula Guedes de Pinho
ZUSAMMENFASSUNG
A method based on headspace solid-phase microextraction (HS-SPME) coupled to gas chromatography-triple quadrupole/mass spectrometry detection (GC-TQ/MS) with a prior derivatization step with O-(2,3,4,5,6-pentafluorobenzyl)hydroxylamine hydrochloride (PFBHA) was developed to quantify carbonyl compounds in different categories of Port wines. Optimal extraction conditions were obtained incubating 2 ml of wine with 2.3 g/l of PFBHA for 10 min and extracted during 20 min at 32 °C. The method was validated for 38 carbonyl compounds (alkanals, alkenals, Strecker aldehydes, dialdehydes, ketones and furan aldehydes) with regard to linearity, repeatability, inter and intra-day precision and accuracy, showing that the method is suitable for the determination of carbonyl compounds in wines. Tawny wines with 'indication of age' (10-40 years old) presented the highest levels of some carbonyl compounds, such as propanal, pentanal, hexanal, Strecker aldehydes, diacetyl, methyl glyoxal, 3-pentanone and 2-furfural, whereas Ruby wines were characterized by the highest amounts of some unidentified compounds.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
SPME-Faseranordnung, 50/30µm DVB/CAR/PDMS, StableFlex (1 cm), needle size 23 ga, Autosampler, pk of 3, plain gray hub
Supelco
SPME-Faseranordnung, 50/30µm DVB/CAR/PDMS, StableFlex (2cm), needle size 24 ga, Autosampler, pk of 3, notched gray hub
Sigma-Aldrich
Benzaldehyd, purified by redistillation, ≥99.5%
Supelco
SPME-Faseranordnung, 50/30µm DVB/CAR/PDMS, StableFlex (1cm), needle size 24 ga, Manual Holder, pk of 3, plain gray hub
Sigma-Aldrich
Diacetyl, 97%
Sigma-Aldrich
Capronaldehyd, 98%
Supelco
SPME-Faseranordnung, 50/30µm DVB/CAR/PDMS, StableFlex (2cm), needle size 23 ga, Autosampler, pk of 3, notched gray hub
Sigma-Aldrich
2-Hexanon, reagent grade, 98%
Supelco
SPME-Faseranordnung Polydimethylsiloxan (PDMS), 100 μm PDMS, Fused Silica (1 cm), needle size 23 ga, Autosampler, pk of 3, red hub
Supelco
SPME-Faseranordnung, 50/30µm DVB/CAR/PDMS, StableFlex (1 cm), needle size 24 ga, Autosampler, pk of 3, plain gray hub
Sigma-Aldrich
Phenylacetaldehyd, ≥90%
Sigma-Aldrich
4-Fluorbenzaldehyd, 98%
Supelco
SPME-Faseranordnung Polydimethylsiloxan (PDMS), 100 μm PDMS, Fused Silica (1 cm), needle size 24 ga, Manual Holder, pk of 3, red hub
Sigma-Aldrich
2-Cyclohexen-1-on, ≥95%
Sigma-Aldrich
Poly(dimethylsiloxan), viscosity 1.0 cSt (25 °C)
Supelco
SPME-Faseranordnung Carboxen/Polydimethylsiloxan (CAR/PDMS), 75 μm CAR/PDMS, Fused Silica (1 cm), needle size 24 ga, Manual Holder, pk of 3, black hub
Supelco
SPME-Faseranordnung Polydimethylsiloxan (PDMS), 100 μm PDMS, Fused Silica (1 cm), needle size 24 ga, Autosampler, pk of 3, red hub
Sigma-Aldrich
trans-2-Hexen-1-al, 98%
Supelco
SPME-Faseranordnung Polydimethylsiloxan/Divinylbenzen (PDMS/DVB), 65 μm PDMS/DVB, Fused Silica (1 cm), needle size 24 ga, Manual Holder, pk of 3, blue hub
Supelco
SPME-Faseranordnung Carboxen/Polydimethylsiloxan (CAR/PDMS), 75 μm CAR/PDMS, Fused Silica (1 cm), needle size 24 ga, Autosampler, pk of 3, black hub
Supelco
SPME-Faseranordnung Carboxen/Polydimethylsiloxan (CAR/PDMS), 85 μm CAR/PDMS, StableFlex (1 cm), needle size 23 ga, Autosampler, pk of 3, light blue hub
Supelco
SPME-Faseranordnung Polydimethylsiloxan/Divinylbenzen (PDMS/DVB), 65 μm PDMS/DVB, StableFlex (1 cm), needle size 23 ga, Autosampler, pk of 3, pink hub
Supelco
SPME-Faseranordnung Polydimethylsiloxan/Divinylbenzen (PDMS/DVB), 65 μm PDMS/DVB, Fused Silica (1 cm), needle size 24 ga, Autosampler, pk of 3, blue hub
Supelco
SPME-Faseranordnung Polydimethylsiloxan/Divinylbenzen (PDMS/DVB), 65 μm PDMS/DVB, StableFlex (1 cm), needle size 24 ga, Manual Holder, pk of 3, pink hub
Sigma-Aldrich
4-Heptanon, 98%
Supelco
SPME-Faseranordnung Carboxen/Polydimethylsiloxan (CAR/PDMS), 85 μm CAR/PDMS, StableFlex (1 cm), needle size 24 ga, Manual Holder, pk of 3, light blue hub
Supelco
SPME-Faseranordnung Polyacrylat (PA), 85 μm Polyacrylate, Fused Silica (1 cm), needle size 24 ga, Autosampler, pk of 3, white hub
Supelco
Tragbarer SPME-Field-Sampler, coating PDMS/DVB
Supelco
SPME-Faseranordnung Polydimethylsiloxan/Divinylbenzen (PDMS/DVB), 65 μm PDMS/DVB, StableFlex (1 cm), needle size 24 ga, Autosampler, pk of 3, pink hub
Supelco
SPME-Faseranordnung Polyacrylat (PA), 85 μm Polyacrylate, Fused Silica (1 cm), needle size 23 ga, Autosampler, pk of 3, white hub