- Tumor HLA?DR expression linked to early intrahepatic recurrence of hepatocellular carcinoma
Tumor HLA?DR expression linked to early intrahepatic recurrence of hepatocellular carcinoma
International Journal of Cancer. Journal International Du Cancer (2005)
Matoba K, et al.