Merck
  • Cyclodextrin functionalized graphene nanosheets with high supramolecular recognition capability: synthesis and host- guest inclusion for enhanced electrochemical performance.
Cyclodextrin functionalized graphene nanosheets with high supramolecular recognition capability: synthesis and host- guest inclusion for enhanced electrochemical performance.
ACS Nano (2010)
Guo, Yujing et al.