- Indium(III) chloride catalyzed three-component coupling reaction: a novel synthesis of 2-substituted aryl(indolyl)kojic acid derivatives as potent antifungal and antibacterial agents.
Indium(III) chloride catalyzed three-component coupling reaction: a novel synthesis of 2-substituted aryl(indolyl)kojic acid derivatives as potent antifungal and antibacterial agents.
Bioorganic & medicinal chemistry letters (2010-11-12)
B V Subba Reddy, M Ramana Reddy, Ch Madan, K Pranay Kumar, M Srinivasa Rao
PMID21067928
ZUSAMMENFASSUNG
Three-component coupling of aldehyde, indole and kojic acid has been achieved using a catalytic amount of InCl(3) under solvent free conditions to produce a novel series of 2-substituted aryl(indolyl)kojic acid derivatives in good yields and with high selectivity. These compounds are found to exhibit potent antifungal properties.