Merck
  • Determination of polar pesticide metabolites in aqueous matrices by LC-MS and investigation of occurring matrix effects.
Determination of polar pesticide metabolites in aqueous matrices by LC-MS and investigation of occurring matrix effects.
Analytical Chemistry (2013)
Kowal, et al.