Merck
Intermediate filaments: structure, dynamics, function, and disease.
Annual review of biochemistry (1994-01-01)
E Fuchs, K Weber
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Vimentin, His tagged human, recombinant, expressed in E. coli, ≥90% (SDS-PAGE), buffered aqueous glycerol solution