Merck
MLK3 is required for mitogen activation of B-Raf, ERK and cell proliferation.
Nature cell biology (2004-07-20)
Deborah N Chadee, John M Kyriakis
ZUSAMMENFASSUNG
The ERK group of mitogen-activated protein kinases (MAPKs) is essential for cell proliferation stimulated by mitogens, oncogenic ras and raf (ref. 1). All MAPKs are activated by MAP3K/MEK/MAPK core pathways and the Raf proto-oncoproteins, especially B-Raf, are ERK-specific MAP3Ks (refs 1-3). Mixed lineage kinase-3 (MLK3) is a MAP3K that was thought to be a cytokine-activated, and comparatively selective, regulator of the JNK group of MAPKs (refs 1, 4-6). Here we report that silencing of mlk3 by RNAi suppressed mitogen and cytokine activation not only of JNK but of ERK and p38 as well. Silencing mlk3 also blocked mitogen-stimulated phosphorylation of B-Raf at Thr 598 and Ser 601, a step required for B-Raf activation. Furthermore, silencing mlk3 prevented serum-stimulated cell proliferation and the proliferation of tumour cells bearing either oncogenic Ki-Ras or loss-of-function neurofibromatosis-1 (NF1) or NF2 mutations. The proliferation of tumour cells containing activating B-raf or raf-1 mutations was unaffected by silencing mlk3. Our results define an unexpected role for MLK3 in mitogen regulation of B-Raf, ERK and cell proliferation.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
MLK3 (1-488), active, GST tagged human, PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution